missing translation for 'onlineSavingsMsg'
Learn More
Learn More
RuCl[(R,R)-Fsdpen](p-cymene) 90.0+%, TCI America™
Specifications
Specifications
| Melting Point | 244°C |
| Quantity | 200 mg |
| Formula Weight | 712.14 |
| Percent Purity | ≥90% (NMR) |
| Physical Form | Crystalline Powder |
| Chemical Name or Material | RuCl[(R,R)-Fsdpen](p-cymene) |
Safety and Handling
| TSCA | No |