missing translation for 'onlineSavingsMsg'
Learn More

Rubidium perchlorate, anhydrous, 99.5% (metals basis), Thermo Scientific Chemicals

Catalog No. p-7048837
Change view
Click to view available options
Quantity:
50 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AA1162818 50 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. AA1162818 Supplier Thermo Scientific Chemicals Supplier No. 01162818

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

CAS: 13510-42-4 | Molecular Formula: ClO4Rb | Molecular Weight: 184.914 g/mol

A perchlorate oxidizing agent. Rubidium perchlorate as chosen because its precipitation provides an excellent separation from strontium and since it is also an accepted weighing form for rubidium.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 13510-42-4
Molecular Formula ClO4Rb
Molecular Weight (g/mol) 184.914
MDL Number MFCD00016294
InChI Key NQYGGOOZLKJKPS-UHFFFAOYSA-M
Synonym rubidium perchlorate, rubidium 1+ ion perchlorate, perchloric acid, rubidium salt, rubidiumperchlorat, perchloric acid, rubidium salt 1:1, clo4.rb, rubidium chlorate vii, acmc-20ajs4, perchloric acid rubidium salt, rubidium 1+ perchlorate ion
PubChem CID 23673707
IUPAC Name rubidium(1+);perchlorate
SMILES [O-]Cl(=O)(=O)=O.[Rb+]

Specifications

Physical Form Crystalline
Quantity 50 g
UN Number UN1481
Solubility Information Soluble in water (180mg/ml at 100°C).
Formula Weight 184.94
Percent Purity 99.5%
Odor Odorless
Refractive Index 1.4701
Sensitivity Hygroscopic
Assay (metals basis)
Chemical Name or Material Rubidium perchlorate, Anhydrous
Show More Show Less
Hazard Category H272-H315-H319-H335
Hazard Statement GHS H Statement
H272-H315-H319-H335
May intensify fire
oxidizer.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement P210-P220-P221-P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P370+P378q-P501c
DOTInformation Transport Hazard Class: 5.1; Packing Group: II; Proper Shipping Name: PERCHLORATES, INORGANIC, N.O.S.
EINECSNumber 236-840-1
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only