missing translation for 'onlineSavingsMsg'
Learn More

Rubidium hydroxide hydrate, 99% (metals basis)

Catalog No. p-7049247
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AA1348106 5 g
AA1348114 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AA1348106 Supplier Thermo Scientific Chemicals Supplier No. 01348106
Only null left

Oxidative chlorination catalyst | CAS: 12026-05-0 | HORb | 102.48 g/mol

Oxidative chlorination catalyst.Rubidium hydroxide hydrate is used in fireworks in place of pure rubidium to get violet color, thereby it prevents the waste of expensive element rubidium. It acts as an intermediate in the synthesis of other rubidium compounds.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 12026-05-0
Molecular Formula HORb
Molecular Weight (g/mol) 102.48
MDL Number MFCD00011194
InChI Key CPRMKOQKXYSDML-UHFFFAOYSA-M
Synonym rubidium hydroxide hydrate, rb.ho.h2o, rubidium hydroxide monohydrate, rubidium hydroxide-water 1/1/1, rubidium 1+ ion hydrate hydroxide, rubidium hydroxide rb oh , hydrate 9ci, rubidium hydroxide hydrate, fused solid-rb
PubChem CID 16212572
IUPAC Name rubidium(1+);hydroxide;hydrate
SMILES [OH-].[Rb+]

Specifications

Color White to Gray
Physical Form Crystalline
Quantity 5 g
Linear Formula RbOH·xH2O
UN Number UN2678
Merck Index 14,8285
Solubility Information Soluble in water and alcohol.
Formula Weight 102.48 (Anhydrous)
Percent Purity 99%
Sensitivity Hygroscopic
Assay (metals basis)
Chemical Name or Material Rubidium hydroxide hydrate
Show More Show Less
Hazard Category H302-H314-H335
Hazard Statement GHS H Statement
H314-H318-H302
Causes severe skin burns and eye damage.
Causes serious eye damage.
Harmful if swallowed.
Precautionary Statement P260-P264b-P270-P271-P280-P303+P361+P353-P304+P340-P305+P351+P338-P310-P330-P331-P363-P501c
DOTInformation Transport Hazard Class: 8; Packing Group: II; Proper Shipping Name: RUBIDIUM HYDROXIDE
EINECSNumber 215-186-0
TSCA Yes
Recommended Storage Ambient temperatures; Store under Nitrogen

RUO – Research Use Only