missing translation for 'onlineSavingsMsg'
Learn More

Rubidium formate hydrate, 99.8% (metals basis), Thermo Scientific Chemicals

Catalog No. p-7049251
Change view
Click to view available options
Quantity:
10 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AA1349909 10 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. AA1349909 Supplier Thermo Scientific Chemicals Supplier No. 01349909

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

CAS: 3495-35-0 | Molecular Formula: CHO2Rb | Molecular Weight: 130.49 g/mol

Rubidium formate hydrate is used in anticoagulants, dental materials, indicators and reagents.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 3495-35-0
Molecular Formula CHO2Rb
Molecular Weight (g/mol) 130.49
MDL Number MFCD00054359
InChI Key ZIMBPNXOLRMVGV-UHFFFAOYSA-M
Synonym rubidium formate, formic acid, rubidium salt, formic acid rubidium salt, 64-18-6 parent, formic acid, rubidium salt 1:1, rubidiumformiat, rubidium 1+ formira, rubidium 1+ formate, rubidium 1+ ion formate, formic acid, rubidiumsalt 1:1
PubChem CID 23673641
IUPAC Name rubidium(1+) formate
SMILES [Rb+].[O-]C=O

Specifications

Color White
Linear Formula RbOOCH·xH2O
Quantity 10 g
Sensitivity Hygroscopic
Solubility Information Miscible in water. Also miscible with ether,acetone,ethyl acetate,methanol and ethanol.
Formula Weight 130.49 (Anhydrous)
Percent Purity 99.8%
Physical Form Crystalline
Assay (metals basis)
Chemical Name or Material Rubidium formate hydrate
Hazard Category H315-H319-H335
Hazard Statement GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only