missing translation for 'onlineSavingsMsg'
Learn More

Rubidium chloride, Puratronic™, 99.975% (metals basis)

CAS: 7791-11-9 | ClRb | 120.92 g/mol

$446.62 - $1714.11

Chemical Identifiers

CAS 11-9-7791
Molecular Formula ClRb
Molecular Weight (g/mol) 120.92
MDL Number MFCD00011187
InChI Key FGDZQCVHDSGLHJ-UHFFFAOYSA-M
Synonym rubidium chloride, rubidium chloride rbcl, rubidium monochloride, rbcl, rubidium 1+ chloride, rubidium 1+ ion chloride, unii-n3shc5273s, rubidiumchloride, rubidium-chlorid, rubidium-chloride
PubChem CID 62683
ChEBI CHEBI:78672
IUPAC Name rubidium(1+) chloride
SMILES [Cl-].[Rb+]
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
AA1085109
View Documents
Thermo Scientific Chemicals
01085109
10 g
Each for $446.62
Only null left
 
AA1085118
View Documents
Thermo Scientific Chemicals
01085118
50 g
Each for $1,714.11
Only null left
 
Description

Description

Rubidium chloride is the mostly used rubidium compound, and finds its applications in various fields ranging from electrochemistry to molecular biology. It is used as a non-invasive biomarker, anergic and apathetic depressives in the field of biology; In addition, appropriate preparation of this salt is used as a radioactive isotope to study the perfusion of heart muscle. In the field of biochemistry, it is used to induce cells to take up DNA.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Chemical Identifiers

11-9-7791
120.92
FGDZQCVHDSGLHJ-UHFFFAOYSA-M
62683
rubidium(1+) chloride
ClRb
MFCD00011187
rubidium chloride, rubidium chloride rbcl, rubidium monochloride, rbcl, rubidium 1+ chloride, rubidium 1+ ion chloride, unii-n3shc5273s, rubidiumchloride, rubidium-chlorid, rubidium-chloride
CHEBI:78672
[Cl-].[Rb+]

Specifications

1390°C
11-9-7791
Crystalline Aggregates
99.975% (metals basis)
MFCD00011187
Soluble in water.
[Cl-].[Rb+]
120.92
CHEBI:78672
Odorless
Hygroscopic
2.8 g/cm3 at 20°C
718°C
White
10 g
ClRb
rubidium chloride, rubidium chloride rbcl, rubidium monochloride, rbcl, rubidium 1+ chloride, rubidium 1+ ion chloride, unii-n3shc5273s, rubidiumchloride, rubidium-chlorid, rubidium-chloride
FGDZQCVHDSGLHJ-UHFFFAOYSA-M
rubidium(1+) chloride
62683
120.92
1.493
White crystals
Rubidium chloride
Videos
Safety and Handling

Safety and Handling

EINECSNumber : 232-240-9

RTECSNumber : VL8575000

TSCA : Yes

Recommended Storage : Ambient temperatures

SDS
missing translation for 'documents'

missing translation for 'documents'

RUO – Research Use Only