missing translation for 'onlineSavingsMsg'
Learn More

Rubidium chloride, Puratronic™, 99.975% (metals basis)

Catalog No. AA1085118
Change view
Click to view available options
Quantity:
10 g
50 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AA1085118 50 g
AA1085109 10 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AA1085118 Supplier Thermo Scientific Chemicals Supplier No. 01085118

CAS: 7791-11-9 | ClRb | 120.92 g/mol

Rubidium chloride is the mostly used rubidium compound, and finds its applications in various fields ranging from electrochemistry to molecular biology. It is used as a non-invasive biomarker, anergic and apathetic depressives in the field of biology; In addition, appropriate preparation of this salt is used as a radioactive isotope to study the perfusion of heart muscle. In the field of biochemistry, it is used to induce cells to take up DNA.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 11-9-7791
Molecular Formula ClRb
Molecular Weight (g/mol) 120.92
MDL Number MFCD00011187
InChI Key FGDZQCVHDSGLHJ-UHFFFAOYSA-M
Synonym rubidium chloride, rubidium chloride rbcl, rubidium monochloride, rbcl, rubidium 1+ chloride, rubidium 1+ ion chloride, unii-n3shc5273s, rubidiumchloride, rubidium-chlorid, rubidium-chloride
PubChem CID 62683
ChEBI CHEBI:78672
IUPAC Name rubidium(1+) chloride
SMILES [Cl-].[Rb+]

Specifications

Boiling Point 1390°C
Melting Point 718°C
Color White
Physical Form Crystalline Aggregates
Quantity 50 g
Assay Percent Range 99.975% (metals basis)
Solubility Information Soluble in water.
Formula Weight 120.92
Odor Odorless
Refractive Index 1.493
Sensitivity Hygroscopic
Appearance White crystals
Density 2.8
Chemical Name or Material Rubidium chloride
Show More Show Less
EINECSNumber 232-240-9
RTECSNumber VL8575000
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only