missing translation for 'onlineSavingsMsg'
Learn More

Rubidium Chloride, Metal min. 99%., MP Biomedicals

Catalog No. p-3547288
Change view
Click to view available options
Quantity:
10 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
ICN10282510 10 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. ICN10282510 Supplier MP Biomedicals Inc Supplier No. 0210282510
Only null left

Solutions may be slightly hazy

Chemical Identifiers

CAS 11-9-7791
Molecular Formula ClRb
Molecular Weight (g/mol) 120.92
MDL Number MFCD00011187
InChI Key FGDZQCVHDSGLHJ-UHFFFAOYSA-M
Synonym rubidium chloride, rubidium chloride rbcl, rubidium monochloride, rbcl, rubidium 1+ chloride, rubidium 1+ ion chloride, unii-n3shc5273s, rubidiumchloride, rubidium-chlorid, rubidium-chloride
PubChem CID 62683
ChEBI CHEBI:78672
IUPAC Name rubidium(1+) chloride
SMILES [Cl-].[Rb+]

Specifications

Quantity 10 g
Assay Percent Range Metal min. 99%.
Formula Weight 120.9
Chemical Name or Material Rubidium Chloride
Recommended Storage Room Temperature