missing translation for 'onlineSavingsMsg'
Learn More

Rubidium chloride, 99.8+%, (trace metal basis)

CAS: 7791-11-9 | ClRb | 120.92 g/mol

$148.06 - $2047.70

Chemical Identifiers

CAS 11-9-7791
Molecular Formula ClRb
Molecular Weight (g/mol) 120.92
MDL Number MFCD00011187
InChI Key FGDZQCVHDSGLHJ-UHFFFAOYSA-M
Synonym rubidium chloride, rubidium chloride rbcl, rubidium monochloride, rbcl, rubidium 1+ chloride, rubidium 1+ ion chloride, unii-n3shc5273s, rubidiumchloride, rubidium-chlorid, rubidium-chloride
PubChem CID 62683
ChEBI CHEBI:78672
SMILES [Cl-].[Rb+]
View More Specs

Products 3
Catalog Number Mfr. No. Quantity Packaging Price Quantity  
Catalog Number Mfr. No. Quantity Packaging Price Quantity  
AC201260100
SDS View Documents
Thermo Scientific Chemicals
201260100
10 g Glass bottle
Each for $148.06
Only null left
 
AC201260500
SDS View Documents
Thermo Scientific Chemicals
201260500
50 g Glass bottle
Each for $495.07
Only null left
 
AC201262500
SDS View Documents
Thermo Scientific Chemicals
201262500
250 g Glass bottle
Each for $2,047.70
Only null left
 
Description

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

For ultra trace analysis
Specifications

Chemical Identifiers

11-9-7791
120.92
FGDZQCVHDSGLHJ-UHFFFAOYSA-M
62683
[Cl-].[Rb+]
ClRb
MFCD00011187
rubidium chloride, rubidium chloride rbcl, rubidium monochloride, rbcl, rubidium 1+ chloride, rubidium 1+ ion chloride, unii-n3shc5273s, rubidiumchloride, rubidium-chlorid, rubidium-chloride
CHEBI:78672

Specifications

1390.0°C
11-9-7791
Crystals
99.8+%
RbCl
15,8417
Solubility in water: 910g/L (20 c). Other solubilities: 1g/0.7mL boiling water, practically insoluble in alcohols
[Cl-].[Rb+]
120.92
CHEBI:78672
≥99.8%
Glass bottle
718.0°C
White
10 g
ClRb
MFCD00011187
rubidium chloride, rubidium chloride rbcl, rubidium monochloride, rbcl, rubidium 1+ chloride, rubidium 1+ ion chloride, unii-n3shc5273s, rubidiumchloride, rubidium-chlorid, rubidium-chloride
FGDZQCVHDSGLHJ-UHFFFAOYSA-M
rubidium(1+) chloride
62683
120.92
Trace Metal Basis
Rubidium chloride
Videos
Safety and Handling

Safety and Handling

EINECSNumber : 232-240-9

SDS
Documents

Documents

RUO – Research Use Only