missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Rubidium carbonate, 99%
Description
Rubidium carbonate is used as a raw materials for preparation of rubidium metal and various rubidium salts, for the manufacturing of special glass, for the manufacturing of high energy density micro cells and crystal scintillation counters. It is also used as a part of a catalyst for preparing short-chain alcohols from feed gas.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications
Rubidium carbonate is used as a raw materials for preparation of rubidium metal and various rubidium salts, for the manufacturing of special glass, for the manufacturing of high energy density micro cells and crystal scintillation counters. It is also used as a part of a catalyst for preparing short-chain alcohols from feed gas.
Solubility
Soluble in water4500 g/L @20°C.
Notes
Hygroscopic. Store under dry inert gas. Store away from bases, water/ moisture.
Rubidium carbonate is used as a raw materials for preparation of rubidium metal and various rubidium salts, for the manufacturing of special glass, for the manufacturing of high energy density micro cells and crystal scintillation counters. It is also used as a part of a catalyst for preparing short-chain alcohols from feed gas.
Solubility
Soluble in water4500 g/L @20°C.
Notes
Hygroscopic. Store under dry inert gas. Store away from bases, water/ moisture.
Specifications
Specifications
| Melting Point | 837°C |
| Quantity | 25 g |
| Odor | Odorless |
| Sensitivity | Very Hygroscopic |
| Linear Formula | Rb2CO3 |
| Solubility Information | Soluble in water4500g/L @20°C. |
| IUPAC Name | rubidium(1+);carbonate |
| Formula Weight | 230.95 |
| Percent Purity | 99% |
| Chemical Name or Material | Rubidium carbonate |
Safety and Handling
| Hazard Category | H315-H319-H335 |
| Hazard Statement | GHS H Statement H315-H319-H335 Causes skin irritation. Causes serious eye irritation. May cause respiratory irritation. |
| Precautionary Statement | P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c |
| EINECSNumber | 209-530-9 |
| RTECSNumber | FG0650000 |
| TSCA | Yes |
| Recommended Storage | Ambient temperatures |
RUO – Research Use Only