missing translation for 'onlineSavingsMsg'
Learn More

Rubbermaid™ WaveBrake™ High-Performance Mopping Systems

Catalog No. 19076528
Change view
Click to view available options
Color:
Yellow
Capacity (English):
35 qt.
44 qt.
Product Type:
High Performance Mopping System
Mop Bucket and Side Press Wringer
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Color Capacity (English) Product Type
19076528 Yellow 35 qt. High Performance Mopping System
19076531 Yellow 44 qt. Mop Bucket and Side Press Wringer
296359294 Yellow 35 qt. -
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. 19076528 Supplier Rubbermaid™ Supplier No. 758088 YEL

Mop bucket/wringer system reduces splashing for safer, more productive cleaning.

  • Large-capacity buckets suit commercial applications
  • 19-076-531 features foot pedal water evacuation; no lifting required
  • Wringers are tested to exceed 50,000 wringing cycles, 58X the life of a traditional wringer
  • Construction is premium tubular steel and structural web molded plastic

Order Info

Choose from systems with side-press or down-press wringers, 35 or 44 quart capacity. Optional Dirty Water Bucket is sold separately.

Specifications

Product Type High Performance Mopping System
Material Plastic
Capacity (English) 35 qt.
Color Yellow
For Use With (Application) For the Largest Commercial Cleaning jobs
Height (English) 36.5 in.