missing translation for 'onlineSavingsMsg'
Learn More
Learn More
GSC Go Science Crazy Rubber Stopper, Size 8, 1 hole 5mm in diameter
Pliable Soft Rubber Stoppers for multiple applications
Supplier: GSC Go Science Crazy RS8110
Description
Dimensions 41mm top x 33mm bottom x 25mm height, 5mm hole size. Durometer is approximately 45. Approximately 9 pieces per poundSpecifications
| 33 mm | |
| Black | |
| 25mm |
| 41 mm | |
| Rubber | |
| 5mm |