Learn More
GSC Go Science Crazy Rubber Stopper, Size 6, Solid (no-hole)
Pliable Soft Rubber Stoppers for multiple applications
Supplier: GSC Go Science Crazy RS6
Description
Dimensions 34mm top x 27mm bottom x 25mm height. Durometer is approximately 45. Approximately 16 pieces per poundGreener Choice Claims
- Environmental benefits include:
- Renewable Energy
Specifications
Black | |
27 mm | |
Rubber |
25mm | |
34 mm |