Learn More
Walter Stern Rubber Policemen with Angled Edge
Convenient scrapers with angle-shaped edge.
Supplier: Walter Stern 201AA
Description
Convenient scrapers with angle-shaped edge
- Scrape precipitate from container
- Hard rubber with angled edges
- Use section of glass rod as handle
Specifications
| Policemen Scraper | |
| Designed to scrape precipitate from container by using section of Glass rod as handle | |
| Rubber |
| 0.3 cm dia. Glass Rod | |
| Angle shaped edge | |
| Glass Rod (sold separately) |