Learn More
GSC Go Science Crazy Rubber Policeman
Catalog No. S24581
Change view
Click to view available options
Material:
Natural Rubber Scraper Attached to Glass Rod
Catalog No. Material
S24581 Natural Rubber Scraper Attached to Glass Rod
1 options
Catalog No. S24581 Supplier GSC Go Science Crazy Supplier No. 183500
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Great tool for working with tissues and cultures.
Our Rubber Policeman is a natural rubber scraper attached to a 5mm x 150mm glass rod. It is used to transfer precipitate or solids onto glass surfaces, or for the transfer of tissue culture cells.
Specifications
Product Type Rubber Policeman
Material Natural Rubber Scraper Attached to Glass Rod
For Use With (Application) Transfer precipitate or solids onto glass surfaces, or transfer tissue culture cells