missing translation for 'onlineSavingsMsg'
Learn More

DWK Life Sciences KIMBLE™ Rubber Adapters with 1/4 in. Hole

Catalog No. 12141417
Change view
Click to view available options
Quantity:
12 Pk.
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
12141417 12 Pk.
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 12141417 Supplier DWK Life Sciences Supplier No. 7739000000

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

KIMBLE™ Rubber Adapters with 1/4 in. Hole feature a 1/4 in. hole in the top for use with plain stem thermometers, gas inlet tubes, and more. Sold in cases of 12, these rubber adapters are a practical solution for common laboratory setups.

  • Rubber adapters with a 1/4 in. hole in the top
  • For use with plain stem thermometers, gas inlet tubes, and similar applications
  • Sold in cases of 12

Specifications

Material Rubber
For Use With Plain stem thermometers, Gas inlet tubes
Includes 1/4 in. hole on top
Color Black
Product Type Thermometer Adapter
Quantity 12 Pk.