missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Ru(OAc)2[(R)-binap], TCI America™
$80.16 - $275.87
Chemical Identifiers
| CAS | 325146-81-4 |
|---|---|
| Molecular Formula | C48H40O4P2Ru |
| Molecular Weight (g/mol) | 843.862 |
| MDL Number | MFCD09753020 |
| InChI Key | XANQJWCSQJDBEA-UHFFFAOYSA-N |
| Synonym | Diacetato[(R)-(+)-2,2′C-bis(diphenylphosphino)-1,1′C-binaphthyl]ruthenium(II) |
| PubChem CID | 118705203 |
| IUPAC Name | acetic acid;[1-(2-diphenylphosphanylnaphthalen-1-yl)naphthalen-2-yl]-diphenylphosphane;ruthenium |
| SMILES | CC(=O)O.CC(=O)O.C1=CC=C(C=C1)P(C2=CC=CC=C2)C3=C(C4=CC=CC=C4C=C3)C5=C(C=CC6=CC=CC=C65)P(C7=CC=CC=C7)C8=CC=CC=C8.[Ru] |
| Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
|---|---|---|---|---|---|---|---|---|---|
| Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
|
R0166200MG
|
TCI America
R0166200MG |
200 mg |
Each for $80.16
|
|
|||||
|
R01661G
|
TCI America
R01661G |
1 g |
Each for $275.87
|
|
|||||
Chemical Identifiers
| 325146-81-4 | |
| 843.862 | |
| XANQJWCSQJDBEA-UHFFFAOYSA-N | |
| 118705203 | |
| CC(=O)O.CC(=O)O.C1=CC=C(C=C1)P(C2=CC=CC=C2)C3=C(C4=CC=CC=C4C=C3)C5=C(C=CC6=CC=CC=C65)P(C7=CC=CC=C7)C8=CC=CC=C8.[Ru] |
| C48H40O4P2Ru | |
| MFCD09753020 | |
| Diacetato[(R)-(+)-2,2′C-bis(diphenylphosphino)-1,1′C-binaphthyl]ruthenium(II) | |
| acetic acid;[1-(2-diphenylphosphanylnaphthalen-1-yl)naphthalen-2-yl]-diphenylphosphane;ruthenium |
Specifications
| 325146-81-4 | |
| C48H40O4P2Ru | |
| 200 mg | |
| XANQJWCSQJDBEA-UHFFFAOYSA-N | |
| acetic acid;[1-(2-diphenylphosphanylnaphthalen-1-yl)naphthalen-2-yl]-diphenylphosphane;ruthenium | |
| 118705203 | |
| Crystalline Powder |
| Brown-Yellow | |
| MFCD09753020 | |
| Diacetato[(R)-(+)-2,2′C-bis(diphenylphosphino)-1,1′C-binaphthyl]ruthenium(II) | |
| CC(=O)O.CC(=O)O.C1=CC=C(C=C1)P(C2=CC=CC=C2)C3=C(C4=CC=CC=C4C=C3)C5=C(C=CC6=CC=CC=C65)P(C7=CC=CC=C7)C8=CC=CC=C8.[Ru] | |
| 843.862 | |
| 841.85 | |
| Ru(OAc)2[(R)-binap] |
Safety and Handling
TSCA : No