Learn More
New England Biolab Ltd RtcB Ligase 25 reactions
Catalog No. 501526393
Encompass
RtcB Ligase from E. coli joins single stranded RNA with a 3' -phosphate or 2' ,3' -cyclic phosphate to another RNA with a 5' -hydroxyl. Ligation requires both GTP and MnCl2 and proceeds through a 3' -guanylate intermediate . For substrates with a 2' ,3' -cyclic phosphate end, hydrolysis to a 3' -phosphate precedes 3' end activation with GMP and ligation.
Catalog No. 501526393 Supplier New England Biolab Ltd Supplier No. M0458S
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
RtcB Ligase from E. coli joins single stranded RNA with a 3' -phosphate or 2' ,3' -cyclic phosphate to another RNA with a 5' -hydroxyl. Ligation requires both GTP and MnCl2 and proceeds through a 3' -guanylate intermediate . For substrates with a 2' ,3' -cyclic phosphate end, hydrolysis to a 3' -phosphate precedes 3' end activation with GMP and ligation.