Learn More
Bullard™ RT Series Tychem™ Respirator Hoods
Description
- Tychem™ QC (quality coated) is a lightweight, supple material coated with polyethylene to provide splash protection against many acids, bases, and other liquid chemicals
- Tychem SL (Saranex™ laminate) is coated with Saranex 23-P to provide protection from an even wider range of chemicals
- RT1/RT3 = Tychem QC
- RT2/RT4 = Tychem SL
- Panoramic view minimizes claustrophobic feeling sometimes experienced in other respirator hoods
- Sport or inflatable neck cuff design provides positive custom seal
- Three-channel air delivery system promotes curvature of hood for better fit
- Long inner and outer bibs for splash protection
- Extra-large acetate lens
- No suspension is required
- Breathing tube connects using a sleeve on the back of the hood and a clamp
Order Info
Lens cover available. (See Cat. No. 17-986-288)
Specifications
Specifications
| Assigned Protection Factor (APF) | 1,000 |
| Material | Tychem QC |
| Product Type | Respirator Hood |
| For Use With (Application) | Industrial environments, petroleum refining, pulp and paper manufacturing, food processing, chemical processing, and pharmaceutical manufacturing |
| For Use With (Equipment) | PA30, EVA, and/or RT Airline respiraor |
| Includes | Sport neck cuff |
| Model | RT3 |