Learn More
Selleck Chemical Llc RSL3 5mg 1219810-16-8 DFS Item
Supplier:  Selleck Chemicals Co Ltd S81555MG
RSL3 is a ferroptosis activator in a VDAC-independent manner,exhibiting selectivity for tumor cells bearing oncogenic RAS. RSL3 binds, inactivates GPX4 and thus mediates GPX4-regulated ferroptosis. *For Research & Development use only. Product is not intended for drug, household, or human consumption.
Catalog No. NC1138195
Only null left
Explore available promotions