Learn More
SignalChem Biotech ROCK2, Active 500ug
Catalog No. 502573276
Encompass
Recombinant mouse ROCK2 (5-554) was expressed using baculovirus in Sf9 insect cells using an N-terminal His tag.
Catalog No. 502573276 Supplier SignalChem Biotech Supplier No. R11M11H500
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Recombinant mouse ROCK2 (5-554) was expressed using baculovirus in Sf9 insect cells using an N-terminal His tag.