Learn More
BINDER™ RS 485 / RS 422 Interface Converter
Description
- 115 V option model
- RS 422 cable set and RS 485 / RS 422 interface converter
- For connection to 10-way plug distributor
Specifications
Specifications
| Description | RS 485 / RS 422 interface converter - 115 V option model, RS 422 cable set and RS 485 / RS 422 interface converter for connection to 10-way plug distributor |
| For Use With (Equipment) | BINDER Chambers Series KBF, KBF P, KBF LQC, KBW, KBWF, KMF, MK, MKF, MKT, MKFT, LIT MK |
| Material | Plastic |
| Type | Interface Converter |
| Voltage | 115 V |