Learn More
RPMI 1640 Medium, Powder, MP Biomedicals
Formulated to support lymphoblastoid cells in suspension culture, but it has since been shown to support a wide variety of cells that are anchorage dependent.
Supplier: MP Biomedicals Inc 091060120
Specifications
| Powder | |
| No Sodium Bicarbonate | |
| 10 x 1 L | |
| 4°C |