missing translation for 'onlineSavingsMsg'
Learn More

Corning™ RPMI 1640 Medium (Mod.) 1X without L-Glutamine

Catalog No. MT17105CV
Change view
Click to view available options
Quantity:
6 x 500 mL
6 x 1 L
With Additives:
HEPES
None
Without Additives:
No Glutamine
No Glutamine, No Phenol Red
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity With Additives Without Additives
MT17105CV 6 x 500 mL None No Glutamine, No Phenol Red
MT15041CV 6 x 500 mL HEPES No Glutamine
MT15040CV 6 x 500 mL None No Glutamine
MT15040CM 6 x 1 L None No Glutamine
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
Catalog No. MT17105CV Supplier Corning™ Supplier No. 17105CV

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Guaranteed low endotoxin levels ≤0.025EU/mL

Named for the site at which it was developed, the Roswell Park Memorial Institute, RPMI 1640 is a modification of McCoy's 5A containing an increased level of inositol.  Examples of cell types for which this medium is appropriate include mammalian myeloma and hybridoma cells such as mouse hybridomas, human leukocytes, and B and T lymphocytes.  RPMI 1640 is also used for suspension cultures or monolayer human leukemic cells.  This formulation does not contain L-glutamine.

Specifications

Form Liquid
Product Type RPMI 1640
Sterility Sterile
With Additives None
Without Additives No Glutamine, No Phenol Red
Quantity 6 x 500 mL
pH 7 to 7.4
Content And Storage 2° to 8°C