missing translation for 'onlineSavingsMsg'
Learn More

MP Biomedicals™ RPMI 1640 Medium

Catalog No. MP091260254
Change view
Click to view available options
pH:
6.9 to 7.2
Content And Storage:
15° to 30°C
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. pH Content And Storage
MP091260254 6.9 to 7.2 15° to 30°C
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. MP091260254 Supplier MP Biomedicals™ Supplier No. 091260254

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

General purpose enriched media with extensive applications for a vast range of mammalian cells including human myeloma, mouse hybridoma, human leukocytes, and B and T lymphocytes

Specifications

Product Type RPMI 1640 Medium
Sterility Sterile
Without Additives No Glutamine
Quantity 500 mL
pH 6.9 to 7.2
Content And Storage 15° to 30°C