missing translation for 'onlineSavingsMsg'
Learn More

Roxithromycin, 97%

Catalog No. AAJ6621309
Change view
Click to view available options
Quantity:
5 g
10 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAJ6621309 10 g
AAJ6621306 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAJ6621309 Supplier Thermo Scientific Chemicals Supplier No. J6621309

Semi-synthetic macrolide antibiotic. | CAS: 80214-83-1 | C41H76N2O15 | 837.06 g/mol

Used to block bacterial protein synthesis. Clinically, used to treat respiratory tract, urinary and soft tissue infections.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 80214-83-1
Molecular Formula C41H76N2O15
Molecular Weight (g/mol) 837.06
MDL Number MFCD00214389
InChI Key RXZBMPWDPOLZGW-KMAKEOJNSA-N
Synonym roxithromycin
PubChem CID 133687104
IUPAC Name (3R,4S,5S,6R,7R,9R,10E,11S,12R,13S,14R)-6-{[4-(dimethylamino)-3-hydroxy-6-methyloxan-2-yl]oxy}-14-ethyl-7,12,13-trihydroxy-4-[(5-hydroxy-4-methoxy-4,6-dimethyloxan-2-yl)oxy]-3,5,7,9,11,13-hexamethyl-10-(2,4,7-trioxa-1-azaoctan-1-ylidene)-1-oxacyclotetradecan-2-one
SMILES CC[C@H]1OC(=O)[C@H](C)[C@@H](OC2CC(C)(OC)C(O)C(C)O2)[C@H](C)[C@@H](OC2OC(C)CC(C2O)N(C)C)[C@](C)(O)C[C@@H](C)\C(=N/OCOCCOC)[C@H](C)[C@@H](O)[C@]1(C)O

Specifications

Melting Point 111°C to 118°C
Flash Point >110°C (230°F)
Packaging Glass bottle
Quantity 10 g
Solubility Information Soluble in water to 18μg/ml
Formula Weight 837.04
Percent Purity 97%
Physical Form Powder
Chemical Name or Material Roxithromycin
Hazard Category H302
Precautionary Statement P264b-P270-P301+P312-P330-P501c
RTECSNumber KF4990000
TSCA No
Recommended Storage Keep cold

RUO – Research Use Only