missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific Chemicals Roxithromycin, 96%

Catalog No. AC459570050
Change view
Click to view available options
Quantity:
5 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AC459570050 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. AC459570050 Supplier Thermo Scientific Chemicals Supplier No. 459570050
Get Up to $1,800 in Chemicals with Purchase
View all promotions

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 80214-83-1
Molecular Formula C41H76N2O15
Molecular Weight (g/mol) 837.06
MDL Number MFCD00214389
InChI Key RXZBMPWDPOLZGW-KMAKEOJNSA-N
Synonym roxithromycin
PubChem CID 133687104
IUPAC Name (3S,5R,6S,7S,9S,10E,11R,12S,13R)-6-[4-(dimethylamino)-3-hydroxy-6-methyloxan-2-yl]oxy-14-ethyl-7,12,13-trihydroxy-4-(5-hydroxy-4-methoxy-4,6-dimethyloxan-2-yl)oxy-10-(2-methoxyethoxymethoxyimino)-3,5,7,9,11,13-hexamethyl-oxacyclotetradecan-2-one
SMILES CC[C@H]1OC(=O)[C@H](C)[C@@H](OC2CC(C)(OC)C(O)C(C)O2)[C@H](C)[C@@H](OC2OC(C)CC(C2O)N(C)C)[C@](C)(O)C[C@@H](C)\C(=N/OCOCCOC)[C@H](C)[C@@H](O)[C@]1(C)O

Specifications

Melting Point 111.0°C to 118.0°C
Flash Point > 110°C
Ignition Residue 0.1% max
Infrared Spectrum Authentic
Assay Percent Range 95% min. (HPLC)
Quantity 5 g
Formula Weight 837.06
Percent Purity 96%
Chemical Name or Material Roxithromycin
Hazard Category GHS Signal Word: Warning
Hazard Statement GHS H Statement
Harmful if swallowed.
Precautionary Statement GHS P Statement
Wash face,hands and any exposed skin thoroughly after handling.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.

RUO – Research Use Only