missing translation for 'onlineSavingsMsg'
Learn More

United Scientific™ Round Coverslips

Catalog No. S000845
Change view
Click to view available options
Diameter (Metric):
12 mm
15 mm
18 mm
20 mm
25 mm
5 product options available for selection
Product selection table with 5 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Diameter (Metric)
S000845 12 mm
S000846 15 mm
S000847 18 mm
S000848 20 mm
S000844 25 mm
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 5 options available.
5 options
Choking Hazard
Catalog No. S000845 Supplier United Scientific™ Supplier No. SQCVRSP12
Only null left
Choking Hazard

The Round Cover Slips are manufactured from soda lime glass.

  • Excellent resistance to chemicals
  • Ideal for neural tissue culture and hard-to-attach cells
  • Thickness ranges from 0.13 to 0.17 mm.
  • Avalible in multiple sizes

Specifications

Product Type Coverslip
Shape Circle
Material Glass
Length (Metric) 12 mm
Width (Metric) 12 mm
Thickness (Metric) 0.13 to 0.17 mm
Diameter (Metric) 12 mm