Learn More
Eppendorf™ Round Buckets for Eppendorf™ Four-Place Swinging Bucket Rotors
For use with 05-400-328 swinging bucket rotor or as replacements for buckets supplied with 05-400-318
Supplier:  Eppendorf™ 022639081
Catalog No. 05400320
Only null left
Description
Description
Accommodate conical and round-bottom tubes from 1.1 to 85mL with appropriate adapters.
Specifications
Specifications
A-4-38 swing bucket rotor for 5702/5702R/5702RH centrifuge
Product Suggestions
Product Suggestions