Learn More
Thermo Scientific™ Round Bucket for Thermo Scientific™ Sorvall™ Stratos Rotor
Catalog No. 75008172
Change view
Click to view available options
Capacity:
18 mL
Catalog No. Capacity
75008172 18 mL
1 options
Catalog No. 75008172 Supplier Thermo Scientific™ Supplier No. 75008172
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
For use with Four-Place Swinging Bucket Rotor 75-003-047
Order Info
Order correct quantity to equip rotor. Aerosol-tight lids (75008173) sold separately. Optional adapters accept assorted tube styles and volumes from 1.5mL to 100mL.
Specifications
For Use With (Equipment) Swing out rotor for Labofuge 400 and 400R centrifuge
Capacity 18 mL
Tube Size 1.5 to 100 mL