Learn More
Fisherbrand™ Round-Bottom Polypropylene Test Tubes With Cap
Catalog No. FB1495910AA
Change view
Click to view available options
Capacity (Metric):
5 mL
14 mL
Sterility:
Non-sterile
Sterile
Graduated:
No
Yes
Packaging:
Blister Pack
Blister Pk.
Bulk
Individually Wrapped
5 product options available for selection
Product selection table with 5 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Capacity (Metric) Sterility Graduated Packaging
FB1495910AA 5 mL Non-sterile No Bulk
FB149561D 5 mL Sterile No Blister Pk.
FB149561J 14 mL Sterile No Blister Pk.
FB149569C 14 mL Sterile Yes Blister Pack
FB149569B 14 mL Sterile Yes Individually Wrapped
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 5 options available.
5 options
Catalog No. FB1495910AA
Fisherbrand™'s round bottom tubes provide reliable containment for various laboratory fluid samples and preparations.
  • Ideal for use in laboratory protocols requiring round bottom tubes
  • Ensures reliable handling of liquid samples
  • Polypropylene tubes are best suited for applications requiring greater thermal and chemical stability, while polystyrene tubes offer high optical clarity
  • Available with and without a snap cap - dual-position snap caps offer both vented and fully closed positions.
  • 3,000 RCF rating for polypropylene tubes, 1,400 RCF rating for polystyrene tubes
  • Dimensions and volumes are approximate
Specifications
Color Translucent
Capacity (Metric) 5 mL
Length (Metric) 75 mm
Material Polypropylene
Disposable Single-use
Packaging Bulk
Sterility Non-sterile
Product Line Fisherbrand
Graduated No
Closure Type Snap Cap
Product Type Test Tube
Show More Show Less