Learn More
Greiner Bio-One Round Bottom Polypropylene Culture Tube with Two-Position Vent Stopper
With graduations.
Supplier:  Greiner Bio-One 187261
 View more versions of this product
Catalog No. 07000215
Only null left
Description
Description
  • Sterile
Specifications
Chemical Identifiers
Dual-position Vent Stopper
Polypropylene
Sterile
Specifications
LDPE
Autoclavable (Without Vent Stopper)
Dual-position Vent Stopper
95 mm
Writing area
800
14 mL
18 mm
Free of detectable DNase/RNase, human DNA and pyrogens
Translucent
Polypropylene
1
Sterile
Product Suggestions
Product Suggestions