missing translation for 'onlineSavingsMsg'
Learn More

IKA™ Round Bottom Flask Inlay

Round bottom flask inlay constructed of anodized aluminum

$322.05 - $805.16

Specifications

Shape Round Bottom
Temperature (Metric) 180°C
View More Specs

Products 5
Catalog Number Mfr. No. Capacity (Metric) Price Quantity  
Catalog Number Mfr. No. Capacity (Metric) Price Quantity  
01183265
View Documents
IKA™
25000637
25 mL
Each for $322.05
Only null left
 
01183266
View Documents
IKA™
25000638
50 mL
Each for $322.05
Only null left
 
01183264
View Documents
IKA™
25000636
10 mL
Each for $322.05
Only null left
 
01183271
View Documents
IKA™
25000641
100 mL
Each for $469.70
Only null left
 
01183275
View Documents
IKA™
25000644
500 mL
Each for $805.16
Only null left
 
Description

Description

  • Round bottom
  • Anodized aluminum construction
  • Temperature resistant
Specifications

Specifications

Round Bottom
180°C
Videos
SDS
missing translation for 'documents'

missing translation for 'documents'