missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Guardian™ CP-7R, Chemical Resistant Rough Finish 7 mil Butyl Gloves
Lightweight butyl gloves with fitted hand styling and a rough finish texture offer protection against a wide range of hazardous chemicals.
$79.17 - $79.17
Specifications
| Certifications/Compliance | ASTM D412 |
|---|---|
| Certifications/Compliance (Europe) | CE |
| Color | Black |
| Hand | Specific |
| Latex Free | Yes |
| Catalog Number | Mfr. No. | Size | Price | Quantity | |||||
|---|---|---|---|---|---|---|---|---|---|
| Catalog Number | Mfr. No. | Size | Price | Quantity | |||||
|
19321119
|
Guardian™
50901 |
X-Small |
Pair for $79.17
|
|
|||||
|
19321120
|
Guardian™
50902 |
Small |
Pair for $79.17
|
|
|||||
|
19321121
|
Guardian™
50903 |
Medium |
Pair for $79.17
|
|
|||||
|
19321122
|
Guardian™
50904 |
Large |
Pair for $79.17
|
|
|||||
|
19321123
|
Guardian™
GGL50905 |
X-Large |
Pair for $79.17
|
|
|||||
Description
- Butyl material resists oxidation, ozone corrosion and abrasions
- Reusable and easy to clean
- Fitted hand style with rough finish texture
- Protects against wide range of hazardous chemicals
- 7 mil thickness, 14 in. length
- Made in the USA
Specifications
| ASTM D412 | |
| Black | |
| Yes | |
| Powdered | |
| 0.17 mm | |
| Gauntlet/Rolled | |
| Unlined | |
| 14 in. |
| CE | |
| Specific | |
| Butyl | |
| 7 mil | |
| Permeation resistance to most acids, aldehydes, alcohols, amides, bases, esters and ketones | |
| Rough | |
| ISO 9001 | |
| 35.5 cm |