missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Guardian™ CP-25R, Chemical Resistant Rough Finish 25 mil Butyl Gloves
Description
- Butyl material resists oxidation, ozone corrosion and abrasions
- Reusable and easy to clean
- Fitted hand style with rough finish texture
- Protects against wide range of hazardous chemicals
- 25 mil thickness, 14 in. length
- Made in the USA
Specifications
Specifications
| Certifications/Compliance | ASTM D412 |
| Certifications/Compliance (Europe) | CE |
| Color | Black |
| Hand | Specific |
| Latex Free | Yes |
| Material | Butyl |
| Powdered | Powdered |
| Size | X-Small |
| Thickness (English) | 25 mil |
| Thickness (Metric) | 0.63 mm |
| Show More |