missing translation for 'onlineSavingsMsg'
Learn More

Diversey™ Roubaix

Catalog No. 17000057
Change view
Click to view available options
Volume (English):
55 gal.
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Volume (English)
17000057 55 gal.
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 17000057 Supplier Diversey™ Supplier No. 100972370

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

General purpose anionic liquid detergent

  • Low foaming properties used for spray application
  • For heavy duty cleaning

Specifications

Product Type Meat Room Cleaner/Degreaser
Sterility Non-sterile
Appearance Colorless
Certifications/Compliance Kosher, Letter of no objection
Container Type Drum
Filtered Non-filtered
For Use With (Application) For heavy duty cleaning in meat rooms
Volume (English) 55 gal.
Volume (Metric) 208.2 L