missing translation for 'onlineSavingsMsg'
Learn More

Rottlerin, 95+%, Thermo Scientific™

 

Supplier:  Thermo Scientific Chemicals 328490100

Quantity 10mg
Packaging Glass bottle

Catalog No. AC328490100


Only null left

Description

Description

This Thermo Scientific brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

Specifications

Chemical Identifiers

82-08-6
516.6
DEZFNHCVIZBHBI-ZHACJKMWSA-N
5281847
(E)-1-[6-[(3-acetyl-2,4,6-trihydroxy-5-methylphenyl)methyl]-5,7-dihydroxy-2,2-dimethylchromen-8-yl]-3-phenylprop-2-en-1-one
C30H28O8
MFCD00017361
rottlerin, mallotoxin, kamalin, unii-e29lp3zmuh, e29lp3zmuh, chembl34241, bmk1-f10, e-1-6-3-acetyl-2,4,6-trihydroxy-5-methylphenyl methyl-5,7-dihydroxy-2,2-dimethyl-2h-1-benzopyran-8-yl-3-phenyl-2-propen-1-one, 1-6-3-acetyl-2,4,6-trihydroxy-5-methyl-benzyl-5,7-dihydroxy-2,2-dimethyl-2h-chromen-8-yl-3-phenyl-propenone, 1-6-3-acetyl-2,4,6-trihydroxy-5-methylphenyl methyl-5,7-dihydroxy-2,2-dimethyl-2h-1-benzopyran-8-yl-3-phenyl-2-propen-1-one
CHEBI:8899
CC1=C(C(=C(C(=C1O)C(=O)C)O)CC2=C(C3=C(C(=C2O)C(=O)C=CC4=CC=CC=C4)OC(C=C3)(C)C)O)O

Specifications

Rottlerin
Authentic
Glass bottle
10mg
15, 8405
Solubility in water: insoluble. Other solubilities: soluble in dmso,chloroform,ethanol
CC1=C(C(=C(C(=C1O)C(=O)C)O)CC2=C(C3=C(C(=C2O)C(=O)C=CC4=CC=CC=C4)OC(C=C3)(C)C)O)O
516.6
CHEBI:8899
98%
82-08-6
98%
C30H28O8
MFCD00017361
rottlerin, mallotoxin, kamalin, unii-e29lp3zmuh, e29lp3zmuh, chembl34241, bmk1-f10, e-1-6-3-acetyl-2,4,6-trihydroxy-5-methylphenyl methyl-5,7-dihydroxy-2,2-dimethyl-2h-1-benzopyran-8-yl-3-phenyl-2-propen-1-one, 1-6-3-acetyl-2,4,6-trihydroxy-5-methyl-benzyl-5,7-dihydroxy-2,2-dimethyl-2h-chromen-8-yl-3-phenyl-propenone, 1-6-3-acetyl-2,4,6-trihydroxy-5-methylphenyl methyl-5,7-dihydroxy-2,2-dimethyl-2h-1-benzopyran-8-yl-3-phenyl-2-propen-1-one
DEZFNHCVIZBHBI-ZHACJKMWSA-N
(E)-1-[6-[(3-acetyl-2,4,6-trihydroxy-5-methylphenyl)methyl]-5,7-dihydroxy-2,2-dimethylchromen-8-yl]-3-phenylprop-2-en-1-one
5281847
516.6
Product Suggestions

Product Suggestions

Videos
Safety and Handling

Safety and Handling

GHS H Statement:
Harmful if swallowed.

GHS P Statement:
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS Signal Word: Warning

EINECSNumber : 201-395-4

SDS
Documents

Documents

Promotions

Promotions