missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Eppendorf™ Rotor for Centrifuge 5910 Ri
Eppendorf Rotor for Centrifuge 5910 Ri-precision in refrigerated centrifugation. Tailor your experiments with varied capacities and speeds, adaptable buckets and adaptors for efficient sample processing. Ideal for temperature-sensitive applications, ensuring accuracy and reproducibility.
Supplier: Eppendorf™ 5895181007
Description
- Precision-engineered for optimal sample separation.
- All fixed-angle rotors, rotor lids, buckets, caps and adapters can be autoclaved (121°C, 20 min.).
- Accessories (adaptors and buckets) further customize your centrifuge setup.
- Aerosol-tight options available.
Specifications
| Rotor | |
| 5263 x g | |
| Centrifuge 5910 Ri | |
| 4 | |
| S-4x400 |
| 4 x 400 mL | |
| 5,100 rpm | |
| Without Buckets | |
| Aerosol-tight caps available |