Learn More
Eppendorf™ Rotor for Centrifuges 5910 Ri and 5920 R
Eppendorf Rotor for Centrifuge 5910 Ri and 5920 R-precision in refrigerated large-capacity centrifugation. Tailor your experiments with varied capacities and speeds, buckets and adaptors for efficient sample processing. Ideal for high-throughput applications, ensuring accuracy and reproducibility.
Supplier: Eppendorf™ 5895121004
Description
- Precision-engineered for optimal sample separation.
- All fixed-angle rotors, rotor lids, buckets, caps and adapters can be autoclaved (121°C, 20 min.).
- Accessories (adaptors and buckets) further customize your centrifuge setup.
- Aerosol-tight options available.
Chemical Identifiers
| Rotor | |
| Centrifuges 5910 Ri and 5920 R |
Specifications
| Rotor | |
| 4,800 x g | |
| Centrifuges 5910 Ri and 5920 R | |
| S-4x750, Aerosol-tight |
| 4 x 750 mL | |
| 4,700 rpm | |
| Without Buckets |