Learn More
Beckman Coulter ROTOR FX241.5P 24X1.5/2.0 EA
Catalog No. 50488417
Encompass
Fixed angle polypropylene rotor FX241.5P (24 X 1.5/2.0 mL) with snap-on lid for microfuge 16
Catalog No. 50488417 Supplier Beckman Coulter Supplier No. A46475
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Fixed angle polypropylene rotor FX241.5P (24 X 1.5/2.0 mL) with snap-on lid for microfuge 16