missing translation for 'onlineSavingsMsg'
Learn More

Eppendorf™ Rotor for Centrifuge 5425 / 5425 R

Catalog No. 13864461
Change view
Click to view available options
Capacity:
18 x 1.5/2.0 mL tubes or spin columns
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Capacity
13864461 18 x 1.5/2.0 mL tubes or spin columns
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 13864461 Supplier Eppendorf™ Supplier No. 5495508007

Eppendorf Rotors for Centrifuge 5425/5425 R, precision meets versatility. Optimize your centrifuge with a range of capacities and speeds. Explore accessory options like adaptors and buckets for tailored sample processing, ensuring optimal results in molecular biology and beyond.

  • Precision-engineered for optimal sample separation
  • All fixed-angle rotors, rotor lids, buckets, caps and adapters can be autoclaved (121°C, 20 min.)
  • Accessories (adaptors and buckets) further customize your centrifuge setup
  • Aerosol-tight and PTFE coated options available

Specifications

Description FA-18x2-Kit, Aerosol-tight - includes rotor lid
Material Aluminum
Type Fixed Angle Rotor
Capacity 18 x 1.5/2.0 mL tubes or spin columns
Max. RCF 18,565 x g
Max. Speed 15,060 rpm
Max. Tube Length 39 mm
Max. Tube Diameter 11 mm
Net Weight 5 lb.
For Use With (Equipment) Centrifuge 5425 / 5425 R
Angle 45°
Certifications/Compliance Certificates of Conformity, Compliance, and Containment Testing
Includes Aerosol-tight QuickLock Lid
No. of Places 18
Biocontainment Aerosol-tight
Autoclavable Autoclavable
Show More Show Less