missing translation for 'onlineSavingsMsg'
Learn More

OHAUS Roto-Rock Genie

Catalog No. 01804401
Change view
Click to view available options
Voltage:
120 V
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Voltage
01804401 120 V
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 01804401 Supplier OHAUS Supplier No. 83041254

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Programable all-in-one benchtop Genie Rocker / Rotator or cycle from Rock to Rotate.

  • Includes Magnetic Platform and Non-slip Rocking Tray.
  • Accessories sold separately
  • Genie clip plate systems designed to handle a variety of vessels from PCR tube, vials, bottels, and sealed bags.
  • Digital set/control for RPM, total time, cycle time.
  • Feature Pause/Resume button allowing the operator to add reagent to a sample during process.

Specifications

Rotating Angle 360°
Speed Range 1 to 35 rpm
Voltage 120 V
Product Type Rocker/Rotator
Model Roto-Rock Genie