Learn More
BUCHI Rotavapor Vapor Duct with Combi-Clip
Description
- Combi-Clip is included for fastening the evaporating flask
Specifications
Specifications
| Material | Borosilicate Glass (3.3) |
| Type | Vapor Duct |
| For Use With (Application) | Glass Assembly V, C, S, E, CR, BY, HP |
| For Use With (Equipment) | Rotavapor R-300 |
| Includes | Combi-Clip |
| Standard Taper Joint Number Center | 24/40 |