Learn More
BUCHI Rotavapor R-300 Rotary Evaporator with Controller and V-300 Pump
Catalog No. 05405205
Change view
Click to view available options
Glassware Assembly Type:
Cold Trap
Vertical
Controller Type:
I-300 Pro Interface
Includes:
Woulff Bottle
Woulff Bottle, F-305 Recirculating Chiller
Woulff Bottle, F-308 Recirculating Chiller
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Glassware Assembly Type Controller Type Includes
05405205 Vertical I-300 Pro Interface Woulff Bottle, F-308 Recirculating Chiller
05405204 Vertical I-300 Pro Interface Woulff Bottle
05001067 Cold Trap I-300 Pro Interface Woulff Bottle
05001074 Vertical I-300 Pro Interface Woulff Bottle, F-305 Recirculating Chiller
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
Catalog No. 05405205 Supplier BUCHI Supplier No. 11SR300252VP22
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
The R-300 meets the highest expectations in convenience and versatility. Its modular design allows extension of the Rotavapor R-300 to a fully integrated system where a central interface regulates each component.
  • Intuitive user interaction
  • Precise operation control for repeatability and safety
  • 1 L or 5 L flask capacity baths with cordless changes and cleaning
  • Up to 220 °C bath heating with 5 L bath
  • Ergonomic handle with manual or electronic lift
  • Choice of P+G coating for added protection of condenser and receiving flask
  • Unattended operation and mobile monitoring available
  • Modular system for control, vacuum, and chiller integration
  • Additional glass assemblies and configurations
  • Cloud services for optimized productivity and automated processes
Specifications
Description Electronic Lift
Capacity (Metric) 3 L
Speed 280 rpm
Temperature Range (Metric) 20°C to 220°C
Includes Woulff Bottle, F-308 Recirculating Chiller
Glassware Assembly Type Vertical
Controller Type I-300 Pro Interface
Type Rotary Evaporator System
Model R-300