Learn More
BUCHI Rotavapor R-300
Buchi Rotavapor R-300 is a convenient and versatile rotary evaporator.
Supplier:  BUCHI 11R300212A102
 View more versions of this product
Catalog No. 05000773
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description
Description
  • Intuitive user interaction
  • Precise operation control for repeatability and safety
  • 1 L or 5 L flask capacity baths with cordless changes and cleaning
  • Up to 220 °C bath heating with 5 L bath
  • Ergonomic handle with manual or electronic lift
  • Choice of P+G coating for added protection of condenser and receiving flask
  • Unattended operation and mobile monitoring available
  • Modular system for control, vacuum, and chiller integration
  • Additional glass assemblies and configurations
  • Cloud services for optimized productivity and automated processes
Specifications
Chemical Identifiers
100/120 V
10 to 280 rpm
SJ 24/40
5 L
Rotary Evaporator
Specifications
100/120 V
10 to 280 rpm
Safety Coating (P+G)
Angled
Rotary Evaporator
SJ 24/40
5 L
20°C to 95°C
Standalone
Diagonal (A)
Electronic Lift System
No
Product Suggestions
Product Suggestions