Learn More
BUCHI Rotavapor R-300
Catalog No. 05000443
Change view
Click to view available options
Condenser Type:
Angled
Cold Trap
Vertical Coil
Temperature Range (Metric):
20°C to 220°C
20°C to 95°C
Includes:
I-300 Interface, V-300 Pump, Woulff Bottle
Standalone
Standalone, Uncoated
Woulff Bottle
Model:
Electronic Lift System
Manual Lift System
7 product options available for selection
Product selection table with 7 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Condenser Type Temperature Range (Metric) Includes Model
05000443 Angled 20°C to 220°C Standalone, Uncoated Manual Lift System
05000449 Angled 20°C to 220°C Standalone Manual Lift System
05000773 Angled 20°C to 95°C Standalone Electronic Lift System
05000935 Angled 20°C to 220°C Standalone Electronic Lift System
05000948 Cold Trap 20°C to 220°C Woulff Bottle Electronic Lift System
05000978 Vertical Coil 20°C to 220°C Woulff Bottle Electronic Lift System
05001055 Cold Trap 20°C to 220°C I-300 Interface, V-300 Pump, Woulff Bottle Electronic Lift System
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 7 options available.
7 options
Catalog No. 05000443 Supplier BUCHI Supplier No. 11R300152A002
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Buchi Rotavapor R-300 is a convenient and versatile rotary evaporator.
  • Intuitive user interaction
  • Precise operation control for repeatability and safety
  • 1 L or 5 L flask capacity baths with cordless changes and cleaning
  • Up to 220 °C bath heating with 5 L bath
  • Ergonomic handle with manual or electronic lift
  • Choice of P+G coating for added protection of condenser and receiving flask
  • Unattended operation and mobile monitoring available
  • Modular system for control, vacuum, and chiller integration
  • Additional glass assemblies and configurations
  • Cloud services for optimized productivity and automated processes
Specifications
Voltage 100/120 V
Capacity (Metric) 5 L
Speed 10 to 280 rpm
Temperature Range (Metric) 20°C to 220°C
Material (Coating) Uncoated
Includes Standalone, Uncoated
Condenser Type Angled
Glassware Assembly Type Diagonal (A)
Type Rotary Evaporator
Model Manual Lift System
Standard Taper Joint SJ 24/40
Timer No
Show More Show Less