Learn More
Chemglass Inc Bump Trap, Fritted, X-Coarse, Rotary Evaporator, 24/40 Top Outer, 24/40 Lower Inner
Catalog No. CG132321
Encompass
Bump Trap, Fritted, X-Coarse, Rotary Evaporator, 24/40 Top Outer, 24/40 Lower Inner. Trap has an extra coarse fritted disc sealed between the lower inner and top outer joint to prevent contents of the flask from entering into the condenser in the event of bumping. All traps have a 24/40 top outer joint and the listed size of the lower inner joint.
Catalog No. CG132321 Supplier Chemglass Inc Supplier No. CG132321
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Bump Trap, Fritted, X-Coarse, Rotary Evaporator, 24/40 Top Outer, 24/40 Lower Inner. Trap has an extra coarse fritted disc sealed between the lower inner and top outer joint to prevent contents of the flask from entering into the condenser in the event of bumping. All traps have a 24/40 top outer joint and the listed size of the lower inner joint.