Learn More
Chemglass Inc Bump Trap, Fritted, X-Coarse, Rotary Evaporator, 24/40 Top Outer, 14/20 Lower Inner
Supplier: Chemglass Inc CG132320
Bump Trap, Fritted, X-Coarse, Rotary Evaporator, 24/40 Top Outer, 14/20 Lower Inner. Trap has an extra coarse fritted disc sealed between the lower inner and top outer joint to prevent contents of the flask from entering into the condenser in the event of bumping. All traps have a 24/40 top outer joint and the listed size of the lower inner joint.