Learn More
Chemglass Life Sciences Bump Trap, 100mL, Rotary Evaporator, 24/40 Top Outer, 24/40 Lower Inner DFS Item
Supplier:  Chemglass Inc CG132003
Bump Trap, 100mL, Rotary Evaporator, 24/40 Top Outer, 24/40 Lower Inner. Used between the vapor tube and evaporation flask on a rotary evaporator to prevent contents of the flask from being drawn into the condenser in the event of bumping. Traps have a 24/40 top outer joint and the listed size of lower inner joint.
Catalog No. CG132003
Only null left
Explore available promotions