Learn More
Thermo Scientific Chemicals Rose Bengal
CAS: 632-69-9 | C20H4Cl4I4Na2O5 | 1019.644 g/mol
Supplier: Thermo Scientific Chemicals A1705314
Description
Rose Bengal is a stain used in the eye drops to identify the eye damage. It finds application in the preparation of foraminifera for microscopic analysis, photosensitizing reactions and as a dye. In synthetic chemistry, it is utilized to convert triplet oxygen to singlet oxygen, which is used in [2+2] cycloadditions with alkenes. Its derivatives have important medical functions and used for the treatment of cancer. It acts as an alternative to phloxine B in Kreyberg's stain for keratin and mucus. As a fluorescent probe, it is used for the determination of NAD binding sites of alcohol dehydrogenase.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
SolubilitySoluble in water.
Notes
Incompatible with strong oxidizing agents.
Specifications
| Rose Bengal | |
| >300°C | |
| MFCD00151169 | |
| 3645857 | |
| 14,8262 | |
| Soluble in water. | |
| C1=C2C(=C(C(=C1I)O)I)OC3=C(C(=C(C=C3C24C5=C(C(=C(C(=C5Cl)Cl)Cl)Cl)C(=O)O4)I)O)I.[Na].[Na] | |
| 1019.644 | |
| 1017.65 |
| 632-69-9 | |
| C20H4Cl4I4Na2O5 | |
| 25 g | |
| Light sensitive | |
| rose bengal disodium salt, r105 sodium, rose bengal sodium, rose-bengal 131 i natrium, food red no. 105, sodium salt, food red color no. 105, sodium salt, rosa bengala sodica 131 i inn-spanish, rose bengale 131 i sodique inn-french, sel disodique de rose bengale iodee 131 i, roseum bengalense 131 i natricum inn-latin | |
| DPBQSRZQYAYDGY-UHFFFAOYSA-N | |
| sodium;4,5,6,7-tetrachloro-3',6'-dihydroxy-2',4',5',7'-tetraiodospiro[2-benzofuran-3,9'-xanthene]-1-one | |
| 87244310 |
Chemical Identifiers
| 632-69-9 | |
| 1019.644 | |
| DPBQSRZQYAYDGY-UHFFFAOYSA-N | |
| 87244310 | |
| C1=C2C(=C(C(=C1I)O)I)OC3=C(C(=C(C=C3C24C5=C(C(=C(C(=C5Cl)Cl)Cl)Cl)C(=O)O4)I)O)I.[Na].[Na] |
| C20H4Cl4I4Na2O5 | |
| MFCD00151169 | |
| rose bengal disodium salt, r105 sodium, rose bengal sodium, rose-bengal 131 i natrium, food red no. 105, sodium salt, food red color no. 105, sodium salt, rosa bengala sodica 131 i inn-spanish, rose bengale 131 i sodique inn-french, sel disodique de rose bengale iodee 131 i, roseum bengalense 131 i natricum inn-latin | |
| sodium;4,5,6,7-tetrachloro-3',6'-dihydroxy-2',4',5',7'-tetraiodospiro[2-benzofuran-3,9'-xanthene]-1-one |
Safety and Handling
GHS H Statement
H315-H319-H335
GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P362-P312-P321-P405-P403+P233-P501a
Warning
EINECSNumber : 211-183-3
RTECSNumber : LM5920000
TSCA : Yes
Recommended Storage : Ambient temperatures
RUO – Research Use Only