Learn More
SignalChem Biotech RIPK2 (K47A), Unactive 100ug
Catalog No. 502573315
Encompass
Recombinant human RIPK2 (K47A) (1-299) was expressed by baculovirus in Sf9 insect cells using an N-terminal GST tag.
Catalog No. 502573315 Supplier SignalChem Biotech Supplier No. R0816G100
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Recombinant human RIPK2 (K47A) (1-299) was expressed by baculovirus in Sf9 insect cells using an N-terminal GST tag.